Learn More
DWK Life Sciences Safety Standard Ground Cone, with collar
Codice prodotto. 15866901 Sfoglia Tutto DWK Life Sciences Prodotti
Click to view available options
Joint Size:
13 mm
17 mm
22 mm
26 mm
32 mm
40 mm
Dimensione della confezione:
pezzi 10
Codice prodotto. 15866901
Marca: DWK Life Sciences 286181402
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
DURAN™ safety ground joints for increased safety requirements.
Securing joints by using our KECK™-products is a well-known and proven method. In order to meet high safety demands, e.g. when dealing with hazardous substances or pressure differences, DURAN™ Safety Joints offer very good protection against accidental loosening of the connection. Furthermore, DURAN™ Safety Joints offer a system to separate jammed cones and sockets. This minimizes the risk of breakage and related threats. The DURAN™ cores with collar are designed for this purpose with an additionally formed glass ring.
  • Core with collar and additionally moulded glass ring prevents unintentional loosening of the ground joint
  • Firmly seated ground joints can be loosened more easily here. The danger of glass breakage and associated risks is thus minimized.
  • Use in equipment for handling hazardous substances or working with pressure differences.g in the high vacuum range
  • Tight closure due to standard ground joint
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifica
Quantità 10/Pk.
Descrizione articolo Use in equipment for handling hazardous substances or working with pressure differences in the high vacuum range; When working under increased safety requirements; Installation in glass apparatus and devices
Tipo With Collar
Joint Size 13 mm
Spessore (sistema metrico) parete 1.5 mm