Learn More
SCAT™ HPLC Safety Caps, V2.0
Sfoglia Tutto SCAT Europe Prodotti
Click to view available options
Materiale:
PTFE
Polypropylene
Dimensioni filo:
GL 28
GL 45
GL/S40
GL45
Include:
1 x PFA 1.6/2.3/3.2 mm OD Fittings, Air Valve
1 x PTFE 4 mm OD Fitting, Air Valve
1 x raccordo PFA 1,6/2,3/3,2 mm OD, valvola aria
2 x PP 1.6/2.3/3.3 mm OD Fittings, Air Valve, Change Indicator, PP Blind Plug, Labeling Field
2x PFA-fitting 1,6/2,3/3,2 mm OD + valvola a
2x raccordi PFA 1,6/2,3/3,2 mm OD, 1x tappo cieco PFA + valvola aria
2x raccordo PFA 1,6/2,3/3,2 mm OD + valvola aria
3 x PP 1.6/2.3/3.3 mm OD Fittings, Air Valve, Change Indicator, 2 x PP Blind Plug, Labeling Field
4 x PFA 1.6/2.3/3.2 mm OD Fittings, Air Valve, Change Indicator, 3 PFA Blind Plug, Labeling Field
Raccordi: 1 x PFA 1,6/2,3/3,2 mm OD, 1 x valvole d'aria
Raccordi: 1 x PFA 1,6/2,3/3,2 mm OD, 9angoli, 1 x valvole d'aria
Raccordi: 2 x PFA 1,6/2,3/3,2 mm OD, 9angoli, 1 x valvole d'aria
Raccordi: 4 x PFA 1,6/2,3/3,2 mm OD, 1 x valvole d'aria
Raccordi: 6 x PFA 1,6/2,3/3,2 mm OD, 1 x valvole d'aria
Raccordo: 2 x PFA 1,6/2,3/3,2 mm OD, 1 x tappo cieco PFA, 1 x valvole di sfiato
Dimensione della confezione:
1 foglio
Pezzo
Descrizione
SCAT™ Safety Caps V2.0 provide maximum safety for your HPLC solvent supply.
- High quality materials ensure maximum safety for lab and staff
- 360° rotatable and up to 200°C autoclavable screw cap for maximum flexibility
- Stable construction for highest safety and easy handling
- Chemically resistant against aggressive organic solvents
- Attractive design and clear arrangement of connections
- Improved design for easy connection of capillaries in various colors
- Free air valve with change indicator included with every Safety Cap
Specifica
Specifica
| Da utilizzare con (apparecchiatura) | Lab Bottle |
| Include | 1 x PTFE 4 mm OD Fitting, Air Valve |
| Materiale | PTFE |
| Dimensioni filo | GL 45 |
| Da utilizzare con (applicazione) | HPLC Systems |
| Tipo | Safety Cap |
| Diametro (sistema metrico) esterno | 4 mm |