Learn More
Saint-Gobain Tubo Tygon™ a elevata purezza, Formula 2475
Click to view available options
Diametro (sistema imperiale) interno:
0.06 in.
0.18 in.
1/2 in.
1/4 in.
3/8 in.
5/16 in.
Diametro (sistema imperiale) esterno:
0.18 in.
0.31 in.
0.37 in.
0.43 in.
1/2 in.
3/4 in.
Spessore (sistema imperiale):
0.06 in.
0.12 in.
Dimensione della confezione:
15m
Pezzo
Descrizione
- Contains no plasticizers that can leach into or contaminate fluid, yet tubing remains flexible
- Virtually no absorption of key fluid constituents into tubing material or fluid adsorption onto tubing walls; fluid alteration and loss is limited
- Ideal for handling sensitive fluids such as pharmaceutical or biological solutions
- Also available braid-reinforced to withstand solutions with elevated working pressure (Contact your Fisher Scientific Customer Service Representative for details)
Order Info
In packs of 50 ft. (15m)
Compliance
Meets USP Class VI, FDA and NSF specifications. Documented biocompatibility to ISO 10993 standards.
Specifica
Specifica
| Certificazione/Conformità | Meets USP Class VI, FDA and NSF specifications. Documented biocompatibility to ISO 10993 standards. |
| Diametro (sistema imperiale) interno | 5/16 in. |
| Diametro (sistema metrico) interno | 7.9 mm |
| Materiale | Tygon |
| Diametro (sistema imperiale) esterno | 0.43 in. |
| Diametro (sistema metrico) esterno | 11 mm |
| Overall Length (English) | 50 ft. |
| Overall Length (Metric) | 15 m |
| Spessore (sistema imperiale) | 0.06 in. |
| Spessore (sistema metrico) | 1.6 mm |