Learn More
Hamilton™ SaltLine™ Luer Lock Syringes
Click to view available options
Volume (sistema metrico):
250 μL
5 mL
10 mL
Dimensione della confezione:
Pezzo
Descrizione
- PTFE luer lock termination is extremely common as it provides the most flexible options for connection of tubing
- Available in a range of volumes
- Applications include microfluidics, drug/nutritional infusions, electrospinning, mass spectrometer calibration, cell feeding and more
Specifica
Specifica
| Autoclavabile | Not Autoclavable |
| Materiale | Borosilicate Glass, PTFE, Stainless Steel, UHMWPE |
| Product Type | SaltLine Luer Lock Gastight Syringe |
| Volume (sistema metrico) | 10 mL |
| Da utilizzare con (apparecchiatura) | Laboratory Syringe Pumps |
| Tipo di collegamento | Luer Lock |