Learn More
Hamilton™ SaltLine™ Luer Lock Syringes
Codice prodotto. p-8543021
Click to view available options
Volume (sistema metrico):
250 μL
5 mL
10 mL
Dimensione della confezione:
Pezzo
Codice prodotto. 10491591
Marca: Hamilton™ 203260
Choose these syringes when a precise flow is critical for almost any application.
  • PTFE luer lock termination is extremely common as it provides the most flexible options for connection of tubing
  • Available in a range of volumes
  • Applications include microfluidics, drug/nutritional infusions, electrospinning, mass spectrometer calibration, cell feeding and more
Specifica
Autoclavabile Not Autoclavable
Materiale Borosilicate Glass, PTFE, UHMWPE
Descrizione Gas sterilizable
Product Type Gastight Syringe
Pressione max 200 psig (13.8 bar)
Temperatura max (sistema metrico) 50°C
Modello 1005
Min. Temperature (Metric) 20°C
Quantità 1/Pk.
Volume (sistema metrico) 5 mL
Tipo di collegamento Luer Lock