Learn More
Thermo Scientific™ Samco™ Exact Volume Transfer Pipettes
Click to view available options
Volume (sistema metrico):
20 μL
40 μL
50 μL
60 μL
80 μL
100 μL
150 μL
200 μL
250 μL
300 μL
400 μL
Lunghezza (sistema metrico):
5.3 cm
5.9 cm
6.5 cm
7.5 cm
8.0 cm
8.3 cm
8.89 cm
9.5 cm
10.19 cm
10.6 cm
Sterilità:
Non-sterile
Descrizione:
100μL Exact Volume Transfer Pipette
150μL Exact Volume Transfer Pipette
20μL Exact Volume Transfer Pipette
200μL Exact Volume Transfer Pipette
300μL Exact Volume Transfer Pipette
40μL Exact Volume Transfer Pipette
400μL Exact Volume Transfer Pipette
50μL Exact Volume Transfer Pipette
60μL Exact Volume Transfer Pipette
80μL Exact Volume Transfer Pipette
Samco Exact Volume Transfer Pipettes; 250μL Exact Volume Transfer Pipettes, Non-sterile (500/Pack, 5000/Case)
Dimensione della confezione:
5000 PCS
pezzi 500
pezzi 5000
Descrizione
Perform precise dispensing of volumes from 20 to 400 μL by using Thermo Scientific™ Samco™ Exact Volume Transfer Pipettes. Samco transfer pipettes are made of shatterproof, nontoxic polyethylene. Their low-affinity surface prevents proteins binding in research applications. They can be frozen in liquid nitrogen, or heat sealed for storage.
- Shatterproof, nontoxic polyethylene
- Nonsterile
- Low affinity surface
- Freezer ready
Specifica
Specifica
| Descrizione | 50μL Exact Volume Transfer Pipette |
| Diametro (sistema metrico) esterno | 2.5 mm |
| Materiale | LDPE |
| Sterilità | Non-sterile |
| Graduato | No |
| Tipo di imballaggio | Bulk 500/Pk 10Pk/Cs 5000/Cs |
| Volume (sistema metrico) | 50 μL |
| Lunghezza (sistema metrico) | 5.9 cm |
| Tipo | Exact Volume Transfer Pipet |
| Monouso | Single-use |