Learn More
Buerkle™ Sampler Mole with Handle and Bar
Codice prodotto. 10563655 Sfoglia Tutto Bürkle Prodotti
Click to view available options
Lunghezza (sistema metrico):
75 cm
Dimensione della confezione:
Pezzo
Please order drill bits separately!
Codice prodotto. 10563655
Marca: Buerkle™ 53501005
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Please order drill bits separately!
Very robust model with specially hardened forged drill bits. For determining soil properties, moisture and structure. The shape of the Mole's drill bits ensures minimum friction when drilling and removing the Mole from the ground. This reduces physical effort to a minimum.
The shape of the Mole's drill bit ensures minimum friction when drilling and removing the Mole from the ground. Seven different drill bits are available, each of which is designed for a particular soil type.
  • Soil sampler
  • Seven different drill bits available
TRUSTED_SUSTAINABILITY
Specifica
Lunghezza (sistema metrico) 75 cm