Learn More
Value Plastics Sanitary to 600 Series Hose Barb Fittings
Sfoglia Tutto Cole-Parmer Prodotti
Click to view available options
Diametro (sistema imperiale) interno:
1 in.
1/2 in.
1/4 in.
3/4 in.
3/8 in.
Include:
1-1/2 in. Sanitary Clamp Size
3/4 in. Sanitary Clamp Size
Dimensione della confezione:
pezzi 100
Beschreibung
Reduce the risk of bacterial growth with smooth surfaces and minimal crevices. Easily transition from sanitary to hose barb connection. Maintain a low profile and ease of assembly with 600 series barb fittings
Spezifikation
Spezifikation
| Certificazione/Conformità | ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP |
| Diametro (sistema imperiale) interno | 1 in. |
| Quantità confezione | 100/Pk. |
| Porta di uscita | Sanitary x 600 Series Barb |
| Metodo di sterilizzazione | Gamma radiation, ethylene oxide (EtO), E-beam, and autoclave |
| Forma | Diritto |
| Include | 1-1/2 in. Sanitary Clamp Size |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur