Learn More
Value Plastics Sanitary to 600 Series Hose Barb Fittings
Codice prodotto. 30054233 Sfoglia Tutto Cole-Parmer Prodotti
Click to view available options
Diametro (sistema imperiale) interno:
1 in.
1/2 in.
1/4 in.
3/4 in.
3/8 in.
Include:
1-1/2 in. Sanitary Clamp Size
3/4 in. Sanitary Clamp Size
Unit Size:
pezzi 100
Product Code. 30054233
Brand: Value Plastics 5011868
Only null left
Add to basket
Add to basket
Sanitary fittings provide a secure connection where preventing contamination is crucial
Reduce the risk of bacterial growth with smooth surfaces and minimal crevices. Easily transition from sanitary to hose barb connection. Maintain a low profile and ease of assembly with 600 series barb fittings
Specifications
Certificazione/Conformità ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP
Diametro (sistema imperiale) interno 3/4 in.
Quantità confezione 100/Pk.
Porta di uscita Sanitary x Hose Barb
Metodo di sterilizzazione Gamma radiation, ethylene oxide (EtO), E-beam, and autoclave
Forma Diritto
Include 1-1/2 in. Sanitary Clamp Size