Learn More
Cole-Parmer Sanitary Clamp Fittings, 45 Degrees Elbow Union, 316 Stainless Steel
Codice prodotto. 30054242 Sfoglia Tutto Cole-Parmer Prodotti
Click to view available options
Include:
1 in. Sanitary Clamp Size
1-1/2 in. Sanitary Clamp Size
2 in. Sanitary Clamp Size
3/4 in. Sanitary Clamp Size
Dimensione della confezione:
Pezzo
Codice prodotto. 30054242
Marca: Cole-Parmer 5011831
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Great for food, beverage, and dairy applications—fittings comply with FDA and 3A standards
Ideal for general-purpose, food, beverage, and pharmaceutical applications. Free technical application support available to assist with tubing selection. 316L stainless steel (SS) offers excellent chemical resistance against corrosive acids and bases
Specifica
Certificazione/Conformità 3 A;FDA
Porta di uscita Sanitary
Forma 45° Elbow
Include 3/4 in. Sanitary Clamp Size