Learn More
Cole-Parmer Sanitary Clamp Fittings, Straight Concentric Reducer, 316 Stainless Steel
Sfoglia Tutto Cole-Parmer Prodotti
Click to view available options
Include:
1 to 1-1/2 in. Sanitary Clamp Size
1 to 3/4 in. Sanitary Clamp Size
2 to 1 in. Sanitary Clamp Size
2 to 1-1/2 in. Sanitary Clamp Size
Dimensione della confezione:
Pezzo
Descrizione
Ideal for general-purpose, food, beverage, and pharmaceutical applications. Free technical application support available to assist with tubing selection. 316L stainless steel (SS) offers excellent chemical resistance against corrosive acids and bases. Common centerline—useful when cavitation is present
Specifica
Specifica
| Certificazione/Conformità | 3 A;FDA |
| Porta di uscita | Sanitary |
| Forma | Diritto |
| Include | 1 to 1-1/2 in. Sanitary Clamp Size |
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.