Learn More
Value Plastics Sanitary Fitting Caps, Polypropylene
Sfoglia Tutto Cole-Parmer Prodotti
Click to view available options
Ingresso:
1-1/2 in. Sanitary Cap
3/4 in. Mini Sanitary Cap
Dimensione della confezione:
pezzi 100
Descrizione
Ideal for capping off fluid transfer system. Sanitary fittings provide a secure connection where preventing contamination is crucial. Constructed of ADCF polypropylene
Specifica
Specifica
| Certificazione/Conformità | ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP |
| Quantità confezione | 100/Pk. |
| Ingresso | 3/4 in. Mini Sanitary Cap |
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.