Learn More
ACROBiosystems SARS-CoV-2 (COVID-19) S1 protein (HV69-70del), His Tag
Marca: ACROBiosystems S1N-C52Hd-1mg
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
It's been reported that Coronavirus can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 76.7 kDa. The protein migrates as 100-140 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifica
QHD43416.1 (HV69-70 del) | |
1 mg | |
Val 16 - Arg 685 | |
Spike,S1 protein,Spike glycoprotein Subunit1,S glycoprotein Subunit1,Spike protein S1,COVID-19 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE. |
76.7 kDa | |
SARS-CoV-2 S1 protein (HV69-70 del), His Tag (S1N-C52Hd) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Arg 685 (Accession # QHD43416.1 (HV69-70 del)). HV69-70 deletion was identified in the SARS-CoV-2 variant (known as 20 B/501 Y.V1, VOC 202012/01, or B0.1.1.7 lineage) which emerged in the United Kindom. | |
-20°C | |
S1 protein | |
SARS-CoV-2 | |
PBS, pH7.4 | |
HEK293 | |
Viral Proteins |