missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS-CoV-2 Membrane Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marca: R&D Systems MAB10690-SP
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Membrane Monoclonal antibody specifically detects Membrane in SARS-CoV-2 samples. It is validated for Immunohistochemistry, Immunocytochemistry
Specifica
| Membrane | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2b |
| Immunohistochemistry, Immunocytochemistry | |
| 1041508 | |
| Immunohistochemistry 3-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| YP_009724369 | |
| E. coli-derived SARS-CoV-2 membrane (M) protein, Met101-His222, Accession # YP_009724369 | |
| 25 μg | |
| Primary | |
| SARS-CoV-2 | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto