Learn More
SARS-CoV-2 Membrane Glycoprotein Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 28882-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The SARS-CoV-2 Membrane Glycoprotein antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of virus SARS-CoV-2 Membrane Glycoprotein. This antibody recognizes virus antigen. The SARS-CoV-2 Membrane Glycoprotein antibody has been validated for the following applications: ELISA analysis.Specifica
| SARS-CoV-2 Membrane Glycoprotein | |
| Polyclonal | |
| Unconjugated | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Virus | |
| Polyclonal Antibody | |
| IgG |
| ELISA | |
| 580 μg/mL | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 43740571 | |
| Store at -20°C. | |
| Liquid |