Learn More
ACROBiosystems SARS-CoV-2 S2 protein (A701V), His Tag
Marca: ACROBiosystems S2N-C52Hc-1mg
Descrizione
The ongoing SARS-COV-2 pandemics has sickened 3.7 million people around the world, claiming over 1,000,000 lives. To deal with this global public health crisis, unprecedented efforts are being made to study the virus, SARS-CoV-2. At present, there are 193 new crown vaccine projects registered with the World Health Organization are being developed. To support these studies, ACRO has developed Mouse Anti-SARS-CoV-2 antibody IgG Serologic Assay kit.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 59.8 kDa.Specifica
59.8 kDa | |
SARS-CoV-2 S2 protein (A701V), His Tag (S2N-C52Hc) is expressed from human 293 cells (HEK293). It contains AA Ser 686 - Pro 1213 (Accession # QHD43416.1 (A701V, F817P, A892P, A899P, A942P, K986P, V987P)). The recombinant protein is expressed from human 293 cells (HEK293) with T4 fibritin trimerization motif and a polyhistidine tag (at the C-terminus. Proline substitutions (F817P, A892P, A899P, A942P, K986P, V987P) are introduced to stabilize the trimeric prefusion state of SARS-CoV-2 S protein.) | |
-20°C | |
S2 protein | |
SARS-CoV-2 | |
HEK293 | |
Viral Proteins |
1 mg | |
Ser 686 - Pro 1213 | |
Spike,S2 protein,Spike glycoprotein Subunit2,S glycoprotein Subunit2,Spike protein S2,COVID-19 | |
Unconjugated | |
His Tag | |
Powder |
Il programma Encompass di Fisher Scientific offre articoli che non fanno parte del nostro portafoglio di distribuzione. Questi prodotti in genere non hanno immagini o descrizioni dettagliate. Tuttavia, ci impegniamo a migliorare la vostra esperienza di acquisto. Si prega di utilizzare il modulo sottostante per fornire un feedback relativo al contenuto di questo prodotto.