missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS-CoV-2 Spike S2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP3-05701
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
SARS-CoV-2 Spike S2 Polyclonal antibody specifically detects SARS-CoV-2 Spike S2 in SARS-CoV-2 samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifica
| SARS-CoV-2 Spike S2 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| COVID-19, S2 protein, SARS-CoV-2, SARS-CoV-2 Spike, Spike glycoprotein Subunit 2, spike protein, Spike protein S2, structural protein, surface glycoprotein | |
| SARS-CoV-2 Spike S2 antibody was raised against a peptide corresponding to 16 amino acids near the carboxy terminus of SARS-CoV-2 Spike S2 glycoprotein. The immunogen is located within 1130-1180 amino acids of SARS-CoV-2 Spike S2 protein. | |
| 50 μg | |
| Primary | |
| SARS-CoV-2 | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, ELISA 1 μg/mL, Immunohistochemistry 0.1ug/mL, Immunocytochemistry/ Immunofluorescence 10ug/mL, Immunohistochemistry-Paraffin | |
| QHD43416 | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 43740568 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto