missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sars Membrane Protein Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Marca: Novus Biologicals NB100-56569
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Sars Membrane Protein Polyclonal specifically detects Sars Membrane Protein in SARS-CoV samples. It is validated for Western Blot.
Specifica
| Sars Membrane Protein | |
| Polyclonal | |
| Unconjugated | |
| PBS containing 0.2% gelatin with 0.05% Sodium Azide | |
| E1 glycoprotein, Matrix glycoprotein, Membrane glycoprotein, SARS coronavirus M protein, SARS coronavirus Membrane protein, SARS CoV, SARS CoV M glycoprotein, SARS CoV Matrix protein, Sars M, Sars Matrix Protein, SARSCoV, SARS-CoV M protein, SARS-CoV Membrane protein, Severe acute respiratory syndrome | |
| The antibody was developed by immunizing rabbits with a synthetic peptide corresponding to amino acids 195-210 (YSRYRIGNYKLNTDH) from the M (Membrane protein) for the Human SARS coronavirus (Genbank accession no. YP_009724393.1) | |
| 0.1 mg | |
| Primary | |
| The NCBI accession number of the SARS matrix protein is NP_828855.1. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 1.0 mg/mL | |
| Western Blot 0.5-2 ug/ml, Immunocytochemistry/Immunofluorescence reported by customer review | |
| P59596 | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| 1489672 | |
| SARS-CoV | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto