missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sars Membrane Protein Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP2-41060-0.025mg
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Sars Membrane Protein Polyclonal specifically detects Sars Membrane Protein in SARS-CoV samples. It is validated for Western Blot, ELISA.
Specifica
| Sars Membrane Protein | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| E1 glycoprotein, Matrix glycoprotein, Membrane glycoprotein, SARS coronavirus M protein, SARS coronavirus Membrane protein, SARS CoV, SARS CoV M glycoprotein, SARS CoV Matrix protein, Sars M, Sars Matrix Protein, SARSCoV, SARS-CoV M protein, SARS-CoV Membrane protein, Severe acute respiratory syndrome | |
| Antibody was raised against a synthetic peptide corresponding to 15 amino acids at the carboxy terminus of the SARS matrix protein. The immunogen is located within the last 50 amino acids of SARS Matrix. Amino Acid Squence: GNYKLNTDHAGSNDN | |
| 0.025 mg | |
| Primary | |
| SARS-CoV | |
| IgG |
| Western Blot, ELISA | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, ELISA 1 μg/mL | |
| P59596 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 1489672 | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto