Learn More
Sartorius Combisart™ Accessories for Colony Counting
Sfoglia Tutto Sartorius Prodotti
Click to view available options
Stile uscita:
Hose Nipple, DN 10
TR 20 x 2mm male thread with DN 24 (≈24mm) hexagonal nut
Dimensione della confezione:
Pezzo
Descrizione
For use with Combisart Individual and Multibranch Systems for Colony Counting (Rep. Cat. Nos. 14-555-800 and 14-555-802).
Specifica
Specifica
| Product Type | Base Support |
| Area di filtraggio | 12.5cm2 |
| Materiale | Stainless Steel |
| Stile uscita | TR 20 x 2mm male thread with DN 24 (≈24mm) hexagonal nut |
| Da utilizzare con (applicazione) | Microbiological analysis or particle count in quality assurance |
| Include | 50mm Frit |