Learn More
Sartorius Support Arm, for Cubis™ High-Capacity Balances
Sfoglia Tutto Sartorius Prodotti
Click to view available options
:
Support Arm for Cubis High Capacity Precision Weighing Modules
Descrizione
For precision weighing modules with 100mg/1g readability and weighing capacity ≥20kg; for raising the MSE, MSU, and MSA display and control units. With the display raised on a support arm, you can set up your balance so that even working with heavy loads is ergonomic and minimally stressful.
Specifica
Specifica
| Da utilizzare con (apparecchiatura) | Cubis High Capacity Balance |
| Tipo | Support Arm |