missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Disposable Stands for Concentrators

Codice prodotto. 10787841 Sfoglia Tutto Sartorius Prodotti
Click to view available options
:
Holds four solvent absorption concentrators
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Codice prodotto. 10787841

Marca: Sartorius VPA002

Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Visualizzare prodotti alternativi

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

3 - 20mL Vivapore™ are ready-to-use static concentrators for fast and affordable concentration of samples. Extremely simple and user-friendly. Stand and pipettes included in packs with 4 and 30 devices. Sample concentration without additional equipment.

Vivapore™ 5 and Vivapore™ 10/20 are a ready-to-use, disposable ultrafiltration devices for samples up to 20 ml initial volume. They are optimally utilized for concentrating macromolecules in clinical specimens such as urine, cerebrospinal fluid (CSF) or other biological solutions. Two different devices are available: Vivapore™ 5 for concentration of 5 ml samples and Vivapore™ 10/20 for concentration of 10 ml samples expandable with an expansion reservoir to 20 ml. With no need for additional equipment, solvent absorption is the most economic and user friendly concentration technique available for clinical applications. Just fill the unit with the solution to be concentrated, wait for the desired concentration level to be achieved and then pipette the concentrated sample from the dead-stop reservoir at the bottom of the concentrator. Vivapore™ static concentrators are offered as separate chambers together with disposable plastic stands for up to four units. Contrary to other, larger products with multiple, adjacent concentration channels, there is no risk of cross-contamination between individual Vivapore™ units. Vivapore™ 5 and 10/20 are frequently used for urine concentration prior to subsequent analysis of Bence-Jones proteins by gel or capillary electrophoresis.

  • Optimal for clinical specimen concentration e.g. urine High-flow 7.5 kDa PES membrane
  • Separate chambers / no cross contamination
  • For up to 20 ml sample volume
TRUSTED_SUSTAINABILITY

Specifica

Product Type Disposable Stand
Autoclavabile Not Autoclavable
Colore White/White
Capacità Solvent Absorption Concentrators
Materiale Stainless Steel
Posizioni 4
Forma Horizontal
Ripiani 4 Tiers
Sovrapponibile Yes
Sterilità Non-sterile
Larghezza (sistema metrico) 7 cm
Altezza (sistema metrico) 6.4 cm
Vedi altri risultati Mostra meno risultati
Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato