Learn More
Sartorius Peristaltic Pump for Vivaflow™ Models
Sfoglia Tutto Sartorius Prodotti
Click to view available options
Portata:
16 to 480 mL/min.
Dimensione della confezione:
Pezzo
Descrizione
Includes
This part (Cat. No. 14-558-339) includes the pump only. The pump head must be purchased separately.
For Vivaflow 50 and 200. Masterflex economy drive variable speed peristaltic pump (115 V).
Specifica
Specifica
| Control Type | Variable Speed |
| Tipo di display | No Display |
| Portata | 16 to 480 mL/min. |
| Motor Speed | 20 to 600 rpm |
| Reversible | Yes |
| Include | Pump only; Pump head is sold separately. |
| Tensione | 115 V |