Learn More
Sartorius Reusable Sterility Testing System: Silicone Adapter
Sfoglia Tutto Sartorius Prodotti
Click to view available options
:
Silicone Adapter; Sterile: No
Descrizione
- With stoppers drilled to fit filter holder base
- Allows sterile filtration system to be used with Sartorius and other stainless-steel and PVC manifolds
Specifica
Specifica
| Monouso | Reusable |
| Da utilizzare con (apparecchiatura) | Reusable Sterility Testing System |
| Materiale | Silicone |
| Sterilità | Non-sterile |
| Tipo | Silicone Adapter |