Learn More
Sartorius Traditional Vacuum Pumps
Sfoglia Tutto Sartorius ProdottiDescrizione
The Microsart™ e.jet vacuum laboratory pump is designed to create sufficient vacuum for filtration and unlike traditional vacuum pumps, it allows simultaneous transfer of the filtered liquid to waste. The Microsart™ e.jet Transfer Pump is ideal for sample filtration in Microbiology achieving a trans membrane pressure below 700 mbar in compliance with ISO 8199 (Water quality - General guidance on the enumeration of micro-organisms by culture). Constant flow rates and a defined maximum vacuum guarantee smooth and reliable filtration. The Microsart™ e.jet Transfer Pump greatly reduces operating complexity and is an ideal accessory for 3-branch and 1-branch manifolds.
The Microsart™ e.jet is designed to pump both gas and liquids, meaning no loss of efficiency or malfunctions from water drawn into the pump head. The vacuum pump series Microsart™ maxi.vac and Microsart™ mini.vac provide up to date technology for daily use in the Microbiology laboratory environment. They are electric neoprene membrane pumps and therefore oil- and maintenance-free with a low noise level. The vacuum produced by these pumps is controlled and can be easily adjusted to your specifications. Thus sensitive cells (e.g. bacteria) are concentrated on the surface of a membrane filter under better conditions, which results in decreased mortality of the cells, higher recovery rates and shorter incubation times.
Specifica
Specifica
| Vuoto max | 100 mbar |
| Requisiti elettrici | 220 V 50 Hz |
| Dimensioni (L x P x A) | 8 x 5.7 x 7.4 in. (203 x 145 x 187 mm) |
| Profondità (sistema metrico) | 18.7 cm |
| Wattage | 80 w |
| Peso (sistema imperiale) | 9.9 lb. |
| Peso (sistema metrico) | 4.5 kg |
| Temperatura (sistema imperiale) max | 104°F |
| Temperatura (sistema metrico) max | 40°C |
| Portata | 20 L/min. |
| Vedi altri risultati |