Learn More
Sartorius Traditional Vacuum Pumps
Codice prodotto. 15972540 Sfoglia Tutto Sartorius Prodotti
Click to view available options
Profondità (sistema metrico):
18.7 cm
22.5 cm
Requisiti elettrici:
110 V 60 Hz
220 V 50 Hz
Dimensione della confezione:
Pezzo
Codice prodotto. 15972540
Marca: Sartorius 16695
Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Visualizzare prodotti alternativi
Neoprene membrane pumps with low noise level, oil- and maintenance-free and is a reliable vacuum
The Microsart™ e.jet vacuum laboratory pump is designed to create sufficient vacuum for filtration and unlike traditional vacuum pumps, it allows simultaneous transfer of the filtered liquid to waste. The Microsart™ e.jet Transfer Pump is ideal for sample filtration in Microbiology achieving a trans membrane pressure below 700 mbar in compliance with ISO 8199 (Water quality - General guidance on the enumeration of micro-organisms by culture). Constant flow rates and a defined maximum vacuum guarantee smooth and reliable filtration. The Microsart™ e.jet Transfer Pump greatly reduces operating complexity and is an ideal accessory for 3-branch and 1-branch manifolds.
The Microsart™ e.jet is designed to pump both gas and liquids, meaning no loss of efficiency or malfunctions from water drawn into the pump head. The vacuum pump series Microsart™ maxi.vac and Microsart™ mini.vac provide up to date technology for daily use in the Microbiology laboratory environment. They are electric neoprene membrane pumps and therefore oil- and maintenance-free with a low noise level. The vacuum produced by these pumps is controlled and can be easily adjusted to your specifications. Thus sensitive cells (e.g. bacteria) are concentrated on the surface of a membrane filter under better conditions, which results in decreased mortality of the cells, higher recovery rates and shorter incubation times.
TRUSTED_SUSTAINABILITY
Specifica
Vuoto max 100 mbar
Amperaggio 0.75 A
Requisiti elettrici 110 V 60 Hz
Dimensioni (L x P x A) 7.3 x 8 x 5.7 in. (18.7 x 20.3 x 14.5 cm)
Profondità (sistema metrico) 18.7 cm
Wattage 80 w
Peso (sistema imperiale) 9.9 lb.
Peso (sistema metrico) 4.5 kg
Temperatura (sistema imperiale) max 104°F
Temperatura (sistema metrico) max 40°C
Portata 20 L/min.
Da utilizzare con Combisart
Frequenza 60 Hz
Overall Height (English) 7.4 in.
Overall Height (Metric) 14.5 cm
Overall Length (English) 8 in.
Lunghezza complessiva (sistema metrico) 203 mm
Overall Width (English) 8 in.
Larghezza complessiva (sistema metrico) 145 mm
Tipo di pompa Vacuum Pump
Applicazioni Single filtration run with individual filter station for microbial enumeration
Tensione 110 V