Learn More
Sartorius Vivapure™ AdenoPACK 20 Kit
BIOPHARMA
Codice prodotto. 10257712 Sfoglia Tutto Sartorius Prodotti
Click to view available options
:
AdenoPACK 20
Codice prodotto. 10257712
Marca: Sartorius VSAVPQ020
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
The optimal kit for construct screening
Vivapure AdenoPACK 20 is the small scale kit in the AdenoPACK series, purifying up to 1 x 1012 viral particles from 20mL cell culture. Especially when it comes to testing new constructs, parallel and fast purifications of different adenoviruses are essential. This kit allows the rapid, simple and affordable spin column based purification of 6 different samples in parallel and bridges a gap in the CsCl density gradient method - for the first time adenovirus can efficiently be purified from less than 100mL cell culture volume.
  • Spin column based kit
  • Easy and fast parallel purification of multiple samples
  • Virus purification based on membrane adsorber technology
  • High-speed process completed in about 1 hour
  • Excellent viral access provides quantitative recoveries
  • Able to process 6 x 20mL cell culture
  • Recovers 1 x 1011 to 1012 viral particles from 20 mL cell culture
For a normal yielding vector, 1 x 15cm culture plate purified using this method yields up to 1 x 1012 viral particles.
TRUSTED_SUSTAINABILITY
Specifica
Concentrazione di endotossine <0.025 EU/mL
Dimensione campione 20 mL of Adenovirus supernatants
Content And Storage -20°C