Learn More
Thermo Scientific™ Savant™ SpeedVac™ Rotors for Medium Capacity Concentrators
Codice prodotto. p-4529004 Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Capacità:
10 Centrifuge Tubes
100 (6 x 50mm) Tubes
100 (8 x 29 mm) Tubes
12 Scintillation Vials
12 Vials
24 Vials
32 Tubes
4 x 100 mL Pear-shaped Flasks
40 Microtubes
40 Tubes
6 x 50 mL Conical Tubes
60 Vials
64 Microtubes
72 Tubes
8 (18 x 100, 17 x 95, 16 x 100 mm) Tubes
Dimensione della confezione:
Pezzo
Codice prodotto. 11758609
Marca: Thermo Scientific™ RH1006
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Select from one of these rotors to fit the type of sample volume and various sample formats for your specific application. Choose from various size of tubes, flasks, mircrowell plates.
SpeedVac Rotors for Medium Capacity Concentrators for use with Savant SpeedVac Concentrators.
Circular Rotors:
  • Easy to use
  • Multi-size tube holes accommodate different sample sizes
  • Require no additional accessories
  • Fit directly into your concentrator
TRUSTED_SUSTAINABILITY
Specifications
Descrizione Glass and Plastic Tubes: 100 (6 x 50mm), Medium capacity models
Materiale Glass (Tube), Plastic (Tube)
Tipo Rotor
Da utilizzare con (apparecchiatura) SPD1030, SPD120, SPD130, SPD140DDA
Capacità 100 (6 x 50mm) Tubes
Modello RH100-6
Sistema rotore Traditional
Dimensioni (L x P) 6 x 50 mm