Learn More
Cole-Parmer™ Scienceware™ Stainless Steel Vibrating Spatula with PTFE Coating
Sfoglia Tutto Cole-Parmer Prodotti
Click to view available options
Lunghezza (sistema imperiale):
8.25 in.
Dimensione della confezione:
Pezzo
Descrizione
- Move the toothed thumbwheel to vibrate the blade and add as little or as much of your material as needed
- Narrow V-shaped blade maneuvers easily into small-mouth containers and tight corners
- Stainless steel blade is also available with a brown PTFE coating
- FDA compliant for direct food contact applications
Specifica
Specifica
| Product Type | Vibrating Spatula |
| Lunghezza (sistema imperiale) | 8.25 in. |
| Lunghezza (sistema metrico) lama | 20.96 cm |
| Materiale | Stainless Steel/PTFE Coated |