missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCN1A Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 28079-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Epithelial sodium channels are amiloride-sensitive members of the Degenerin/epithelial sodium channel (Deg/ENaC) superfamily of ion channels. Members of this superfamily of ion channels share organizational similarity in that they all possess two short intracellular amino and carboxyl termini, two short membrane spanning segments, and a large extracellular loop with a conserved cysteine-rich region. There are three homologous isoforms of the ENaC (alpha, beta, and gamma) protein. ENaC in the kidney, lung, and colon plays an essential role in trans-epithelial sodium and fluid balance. ENaC also mediates aldosterone-dependent sodium reabsorption in the distal nephron of the kidney, thus regulating blood pressure. ENaC is thought to be regulated, in part, through association with the cystic fibrosis transmembrane conductance regulator (CFTR) chloride ion channel. Gain-of-function mutations in beta- or gamma-ENaC can cause severe arterial hypertension (Liddels syndrome) and loss-of-function mutations in alpha- or beta-ENaC causes pseudohypoaldosteronism (PHA-1).Specifica
| SCN1A | |
| Polyclonal | |
| Unconjugated | |
| SCN1A | |
| FEB3, GEFSP2, HBSCI, NAC1, Nav1.1, SCN1, SCN1A, SMEI | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 6323, 81574 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P04774, P35498 | |
| SCN1A | |
| SCN1A Fusion Protein Ag25942 | |
| 20 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto