Learn More
Ohaus™ Screw Cap, 250 mL Rack Adaptor, Frontier™ R-S4x250/4MI Swing Out Rotors
Sfoglia Tutto Ohaus Prodotti
Click to view available options
:
For use with 250 mL Rack Adaptor for Frontier™ R-S4x250/4MI Swing Out Rotors, O-rings
Descrizione
- Suitable for use with 250 mL Rack Adaptor for Frontier™ R-S4x250/4MI Swing Out Rotors
- Requires 250 mL bucket cap O-rings
Specifica
Specifica
| Da utilizzare con (apparecchiatura) | 250 mL Rack Adaptor , Frontier, R-S4x250/4MI Swing Out Rotors, O-rings |
| Descrizione | Screw Cap, Rack of 250ml Rotor, 2/pk |