Learn More
DWK Life Sciences Screw Cap closed, with seal for safety standard ground sockets
Sfoglia Tutto DWK Life Sciences Prodotti
Click to view available options
Joint Size:
NS 14/23
NS 19/26
NS 24/29
NS 29/32
NS 34/35
NS 45/40
Dimensione della confezione:
pezzi 10
Descrizione
With closed screw caps, devices made of DURAN™ safety grinding can be securely closed after use. The PTFE-coated silicone seal provides maximum chemical resistance.
- Secure closure of the safety ground joints prevents unintentional opening of the ground joint
- PTFE-coated silicone gasket for maximum chemical resistance
Specifica
Specifica
| Quantità | 10/Pk. |
| Descrizione articolo | Suitable for sealing security grinding |
| Joint Size | NS 14/23 |