Learn More
Eppendorf™ Sealing Mat for DWP96/2000 Deepwell Plate
Sfoglia Tutto Eppendorf Prodotti
Click to view available options
:
For use with DWP 96/2000 Deep Well Plate
Descrizione
- Storage film and foil-glued seals for simple and dependable sealing during sample storage
- Heat sealing rilm and foil for continuous locking with the best evaporation protection
- Foil products are made of aluminum, can be pierced, and protect light-sensitive samples
- Film products are transparent and provide protection against unwanted punctures
- Eppendorf plate lid: stable, flexible protection of samples during short-term storage
- Autoclavable
Specifica
Specifica
| Descrizione | PCR clean |
| N. per confezione | 500/Pk., 5 Bags x 10 Pieces |
| Plate Compatibility | 96/2000 Deep Well Plate |