Learn More
DWK Life Sciences Sealing ring, for polished spherical joints (silicone + FEP)
Sfoglia Tutto DWK Life Sciences Prodotti
Click to view available options
Descrizione:
19 NS
29 NS
35 NS
41 NS
64 NS
S 13 NS
Dimensione della confezione:
pezzi 10
Descrizione
Non-sanded DURAN™ spherical ground joints only seal via the sealing ring. FEP-coated silicone rings are used where high chemical resistance is required.
- Due to the material properties, it can be used for work with high demands on the chemical resistance of the sealing ring.
- Silicone and FEP sealing ring
Specifica
Specifica
| Descrizione | 29 NS |
| Autoclavabile | Not Autoclavable |
| Materiale | Silicone, FEP |
| Da utilizzare con (applicazione) | Sealing of non-sanded spherical ground joints |
| Quantità confezione | 10/Pk. |