Learn More
DWK Life Sciences Sealing Ring, for polished spherical joints (VITON)
Sfoglia Tutto DWK Life Sciences Prodotti
Click to view available options
Joint Size:
13 NS
19 NS
29 NS
64 NS
Dimensione della confezione:
pezzi 10
Descrizione
Ball joints without ground joints only seal via the sealing ring. VITON sealing rings are used in vacuum applications.
- Due to the material properties, it can be used for work with high demands on the chemical resistance of the sealing ring.
- Sealing ring made of VITON
Specifica
Specifica
| Da utilizzare con (applicazione) | Sealing of non-sanded spherical ground joints |
| Joint Size | 64 NS |
| Quantità | 10/Pk. |
| Materiale | Viton |