Learn More
SEC11A Mouse anti-Human, Unconjugated, Clone: 2B11G12, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 67379-1-IG-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The SEC11A antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SEC11A. This antibody recognizes human, mouse, rat antigen. The SEC11A antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifica
| SEC11A | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, Immunohistochemistry, ELISA | |
| 2B11G12 | |
| Unconjugated | |
| 20 μL | |
| 23478 | |
| IgG2b |