missing translation for 'onlineSavingsMsg'
Learn More

Novex™ SelectFX™ Alexa Fluor™ 488 Peroxisome Labeling Kit, for fixed cells

Codice prodotto. 10605393
Click to view available options
Quantità:
1 Kit
Dimensione della confezione:
5mL
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Codice prodotto. 10605393

Marca: Novex™ S34201

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Visualizzare prodotti alternativi

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

SelectFX™ Alexa Fluor™ 488 Peroxisome Labeling Kit, for fixed cells

The SelectFX Alexa Fluor 488 Peroxisome Labeling Kit provides all the reagents needed to label the peroxisomes in fixed cells and includes cell fixative and permeabilization reagents. For detection, the kit uses a primary antibody directed against peroxisomal membrane protein 70 (PMP 70), which is a high-abundance integral-membrane component of peroxisomes, and an Alexa Fluor 488 dye-labeled secondary antibody. The fluorescence can be observed using the same optical filters used for fluorescein.

Applications: peroxisome staining
Product size: 1 kit pack size

Peroxisomes are single membrane-bound vesicles that are found in most eukaryotic cells. Their chief function is to enzymatically oxidize fatty acids and to subsequently catalyze the breakdown of H2O2, a by-product of fatty acid degradation. Recently, interest in peroxisomes has increased especially in studies related to peroxisomal origin and maintenance. Morphological abnormalities in peroxisomes related to disease states and diet have also been on the forefront of current research. The SelectFX Alexa Fluor 488 Peroxisome Labeling Kit provides all the reagents needed to label the peroxisomes in fixed cells, and includes cell fixative and permeabilization reagents. For detection, the kit uses an antibody directed against peroxisomal membrane protein 70 (PMP 70), which is a high-abundance integral-membrane component of peroxisomes. PMP 70 is significantly induced by administration of hypolipidemic agents in parallel with peroxisome proliferation and the induction of peroxisomal fatty acid beta-oxidation enzymes.
TRUSTED_SUSTAINABILITY

Specifica

Formato Slide(s)
Da utilizzare con (apparecchiatura) Fluorescence Microscope
Quantità 1 Kit
Metodo di rilevamento Fluorescence
Linea di prodotti Alexa Fluor, SELECTFX
Shipping Condition Room Temperature
Label Type Alexa Fluor Dyes
Product Type Peroxisome Label

For Research Use Only. Not for use in diagnostic procedures.

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato