Learn More
Selenium powder, -200 mesh, 99.999% (metals basis)
Product Code. 11373248
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11373248 50 g 50g
11363248 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11373248 Supplier Thermo Scientific Alfa Aesar Supplier No. 036208.18
 Restricted Item
In stock
Add to basket
CAS: 7782-49-2 | Se | 78.97 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7782-49-2
Molecular Formula Se
Molecular Weight (g/mol) 78.97
MDL Number MFCD00134090 MFCD00011224
InChI Key BUGBHKTXTAQXES-UHFFFAOYSA-N
Synonym selenium, elemental, selen, hydride, atom, elemental, selenio, vandex, alloy, and compounds, gray
PubChem CID 6326970
ChEBI CHEBI:27568
IUPAC Name selenium
SMILES [Se]
Specifications
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.999% (metals basis)
UN Number UN3077
Percent Purity 99.999%
Assay (metals basis)
Chemical Name or Material Selenium