Learn More
Keystone Cleanroom Products Self-Sealing 1422A Tyvek™ Autoclave Bag with Steam Indicator, 13 to 18 in. wide
Sfoglia Tutto Keystone Cleanroom Products Prodotti
Click to view available options
Lunghezza (sistema imperiale):
14 in.
16 in.
18 in.
20 in.
24in.
26 in.
30 in.
32 in.
36 in.
38 in.
40 in.
43 in.
44 in.
48 in.
54 in.
70 in.
Larghezza (sistema imperiale):
14 in.
15 in.
16 in.
18 in.
Dimensione della confezione:
pezzi 250
Descrizione
- Designed for steam sterilization
- Tyvek™ on both sides for maximum breathability during sterilization
- Integrated steam indicator
- Recommended for sterilization of pharmaceutical equipment
- Manufactured and packaged in controlled cleanroom environment
- Eliminates need to cut material and the use of autoclave tape
- Self-sealing closure is fast and consistent for equipment preparation area
- Creates secondary level of protection during aseptic presentation of equipment
- Resists tearing and punctures
- Superior microbial barrier
- Certificates of conformance sent with shipment
Specifica
Specifica
| Autoclavabile | Autoclavable |
| Capacità (sistema imperiale) | 480 oz. |
| Capacità (sistema metrico) | 14.2 L |
| Certificazione/Conformità | COC |
| Tipo di chiusura | Self-seal |
| Colore | White |
| Monouso | Single-use |
| Da utilizzare con (applicazione) | Steam Sterilization |
| Da utilizzare con (apparecchiatura) | Autoclave Bag |
| Include | Steam Sterilization Indicator |
| Vedi altri risultati |