Learn More
Thermo Scientific™ Septa
Codice prodotto. 15264543 Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Descrizione:
BTO
Marathon
Setto BTO; 17 mm
TR-Blue
TR-Green
Diametro (sistema metrico) interno:
9 mm
11 mm
11.5 mm
12.7 mm
17 mm
Temperatura (sistema metrico) max:
200°C to 250°C
300°C
350°C
400°C
Quantità:
48 (Blister Pk.)
50 (Blister Pk.)
50 (Glass Jar)
Dimensione della confezione:
Pezzo
pezzi 48
pezzi 50
Codice prodotto. 15264543
Marca: Thermo Scientific™ 313B3233
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Use low bleed, robust BTO Septa for demanding GC-MS applications.
Ensure optimal performance of your GC instrument with bleed and temperature optimized Thermo Scientific™ BTO and TR-Green Septa. Made of low-bleed silicone, these septa have excellent mechanical properties, are ideal for demanding GC-MS applications, and may be used reliably up to 400 °C.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifica
Tipo Low Coring Septum
Descrizione TR-Blue
Temperatura (sistema metrico) max 200°C to 250°C
Da utilizzare con (apparecchiatura) Trace and Focus GC Instruments
Materiale Silicone
Diametro (sistema metrico) interno 11 mm
Quantità 50 (Glass Jar)