Learn More
3M™ Series G2001 Hard Hat with Leather Sweatband
Click to view available options
Colore:
Blue
White
Yellow
Dimensione della confezione:
pezzi 20
Descrizione
- A safety helmet with excellent ventilation a good field of vision and one of few lightweight helmets available
- 3M™ Uvicator™ sensor disc tells you when your hard hat has been over exposed to UV- radiation and its time to replace it
- Extended edge on the back of the helmet provides extra protection for the sensitive neck area
- Flat surface for printing a logotype, brand name, etc
- Easy to replace sweatband for comfort and hygiene
- Electrical insulation to 440Vac
Specifica
Specifica
| Colore | Yellow |
| Include | Leather Sweatband |
| Materiale | UV-stabilized ABS |
| Product Type | Helmet |
| Size | 54 to 62 cm |
| Peso (sistema metrico) | 340 g |
| Ventilato | Not Ventilated |