missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Serine racemase Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA564916
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Immunogen sequence: QVPLVDALVVP VGGGGMLAGI AITVKALKPS VKVYAAEPSN ADDCYQSKLK GKLMPNLYPP ETIADGVKSS IGLNTWPIIR DLVDDIFTVT EDEIKCATQ Highest antigen sequence identity to the following orthologs - mouse 93%, rat 92%.
SRR (serine racemase) catalyzes the synthesis of D-serine from L-serine.
Specifica
| Serine racemase | |
| Polyclonal | |
| Unconjugated | |
| Srr | |
| D-serine ammonia-lyase; D-serine dehydratase; ILV1; ISO1; L-serine ammonia-lyase; L-serine dehydratase; Serine racemase; SRR; Srs | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 63826 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9GZT4 | |
| Srr | |
| Recombinant Human SRR. Recombinant protein control fragment (Product #RP-105710). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Spot an opportunity for improvement?Share a Content Correction