missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serotonin Monoclonal Antibody (5HT-H209), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA512111
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
MA5-12111 targets Serotonin in IHC (P) applications and shows reactivity with Human samples. The MA5-12111 immunogen is 5-Hydroxytryptamine hydrochloride (3-(2-aminoethyl)-5-hydroxyindole;serotonin).
Serotonin (also known as 5-hydroxytryptamne or 5-HT) is a biochemical messenger and neurotransmitter synthesized from L-tryptophan. Serotonin has a range of functions, including regulation of gastric motility, and may be related to some psychiatric disorders including schizophrenia and depression. Serotoinin is widely distributed in the central and peripheral nervous system, and also in some non-nueral tissues such as platelets.
Specifica
| Serotonin | |
| Monoclonal | |
| Conc. Not Determined | |
| tissue culture supernatant with 0.09% sodium azide | |
| Mouse | |
| 500 μL | |
| Primary | |
| 4° C | |
| Liquid | |
| Chemical |
| Immunohistochemistry (Paraffin) | |
| 5HT-H209 | |
| Unconjugated | |
| 5-HT; 5-hydroxytryptamne | |
| 5-Hydroxytryptamine hydrochloride (3-(2-aminoethyl)-5-hydroxyindole;serotonin) | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG1 κ |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto