missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One VACUETTE™ CAT Serum Clot Activator Blood Collection Tubes
Click to view available options
Capacità (sistema metrico):
4 mL
6 mL
9 mL
Dimensione della confezione:
pezzi 1200
pezzi 50
Descrizione
- VACUETTE™ Blood Collection Tubes are evacuated for easy, safe sample collection. The amount of vacuum in each tube corresponds to the tube’s blood volume specification and helps ensure the correct blood-to-additive ratio.
- Injection molded from polyethylene terephthalate (PET) plastic, VACUETTE™ Blood Collection Tubes are highly transparent and virtually unbreakable.
- VACUETTE™ Serum tubes include a clot-activating additive – micronized silica particles – that is spray-dried on the interior tube wall for even disbursement and optimal blood/additive mixing.
- Optimal fill marks on every tube label indicate the proper draw volume for correct blood-to-additive ratios (within ±10% of the stated nominal volume in accordance with ISO and CLSI).
- Featuring red VACUETTE™ SAFETY Caps, designed to minimize the spraying of aerosols during decapping and color-coded to aid first-glance tube recognition.
- Sterile and latex-free.
Specifica
Specifica
| Colore | Transparent |
| Product Type | Blood Collection Tube |
| Additivo | Clot Activator |
| Materiale | PET |
| Sterilità | Sterile |
| Tipo di chiusura | Non-ridged Pull Cap |
| Capacità (sistema metrico) | 9 mL |
| Da utilizzare con (apparecchiatura) | VACUETTE™ Blood Collection Needles |
| Diametro (sistema metrico) | 13 mm |
| Colore chiusura | Red with Black Ring |
| Vedi altri risultati |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto