Learn More
Greiner Bio-One VACUETTE™ CAT Serum Separator Clot Activator Blood Collection Tubes
Codice prodotto. p-6962057
Click to view available options
Capacità (sistema metrico):
3.5 mL
Colore chiusura:
Gold with Gold Ring
Dimensioni provetta:
13 x 75 mm
Dimensione della confezione:
pezzi 50
Codice prodotto. 15452540
Marca: Greiner Bio-One 454228
Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Visualizzare prodotti alternativi
VACUETTE™ CAT Serum Separator Clot Activator tubes are evacuated blood collection tubes used for serum determinations. They contain clot-activating micronized silica particles and a gel separator that, once centrifuged, forms a stable barrier separating the serum from fibrin and cells.
  • VACUETTE™ Blood Collection Tubes are evacuated for easy, safe sample collection. The amount of vacuum in each tube corresponds to the tube’s blood volume specification and helps ensure the correct blood-to-additive ratio.
  • Injection molded from polyethylene terephthalate (PET) plastic, VACUETTE™ Blood Collection Tubes are highly transparent and virtually unbreakable.
  • VACUETTE™ Serum tubes include a clot-activating additive – micronized silica particles – that is spray-dried on the interior tube wall for even disbursement and optimal blood/additive mixing.
  • Serum tubes with gel contain a barrier gel to separate the serum from the fibrin and cells. During centrifugation, the barrier gel moves upward to the serum/clot interface, where it forms a stable barrier. This barrier separates the serum from fibrin and cells.
  • Optimal fill marks on every tube label indicate the proper draw volume for correct blood-to-additive ratios (within ±10% of the stated nominal volume in accordance with ISO and CLSI).
  • Featuring red VACUETTE™ SAFETY Caps, designed to minimize the spraying of aerosols during decapping and color-coded to aid first-glance tube recognition.
  • Sterile and latex-free.
TRUSTED_SUSTAINABILITY
Specifica
Colore Transparent
Product Type Blood Collection Tube
Additivo Clot Activator and Gel
Materiale PET
Sterilità Sterile
Tipo di chiusura Non-ridged Pull Cap
Capacità (sistema metrico) 3.5 mL
Diametro (sistema metrico) 13 mm
Colore chiusura Gold with Gold Ring
Dimensioni provetta 13 x 75 mm
Lunghezza (sistema metrico) 75 mm