missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SF20/MYDGF Antibody (1009420) [DyLight 650], Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals FAB1147W
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
SF20/MYDGF Monoclonal antibody specifically detects SF20/MYDGF in Human samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), CyTOF
Specifica
| SF20/MYDGF | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
| 1009420 | |
| Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| C19orf10, chromosome 19 open reading frame 10, EUROIMAGE1875335, IL25, IL27, IL27w, interleukin 27 working designation, interleukin-25, Ly6elg, MYDGF, myeloid-derived growth factor, R33729_1, SF20, Stromal cell-derived growth factor SF20, UPF0556 protein C19orf10 | |
| Chinese hamster ovary cell line CHO-derived recombinant human SF20/MYDGF, Met1-Leu173, Accession # Q969H8 | |
| 0.1 mL | |
| Biologically Active Proteins, Innate Immunity | |
| 56005 | |
| Store at 4°C in the dark. | |
| IgG1 |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto