Learn More
Trajan™ Diamond GC Needles for 100 to 500μL Manual Syringes
Codice prodotto. 15298583
Click to view available options
Calibro dell'ago:
23 G
25 G
Include:
Ultra Smooth Coating
Ultra Smooth Hydrophilic Coating
Ultra Smooth Hydrophobic Coating
Dimensione della confezione:
Pezzo
Codice prodotto. 15298583
Marca: Trajan™ 0355312
Removable needle design ensures carryover levels are reduced even lower than the best fixed needle syringes available. Trajan Scientific and Medical™ Diamond GC Needles for 100 to 500μL Manual Syringes are MS ready and designed to improve the sensitivity of your MS results.
Improve the sensitivity of your MS analysis:
  • Lowest syringe carryover
  • Reduced non-specific sample interaction and increased analyte recovery
  • Ultra smooth removable needles with choice of surface treatment.
Specifica
Calibro dell'ago 25 G
Diametro (sistema imperiale) interno 0.007 in.
Diametro (sistema imperiale) esterno 0.0196 in.
Diametro (sistema metrico) interno 0.2 mm
Diametro (sistema metrico) esterno 0.49 mm
Include Ultra Smooth Hydrophilic Coating
Lunghezza (sistema imperiale) ago 2 in.
Product Type GC Needle
Stile del punto dell'ago Bevel
Da utilizzare con (apparecchiatura) Manual Syringe (100 to 500μL)