Learn More
Trajan™ Diamond MS Manual Syringes
Click to view available options
Volume (sistema metrico):
10 μL
25 μL
50 μL
100 μL
250 μL
500 μL
Modello:
100R-GT-MS1
10R-GT-MS1
250R-GT-MS1
25R-GT-MS1
500R-GT-MS1
50R-GT-MS1
Dimensione della confezione:
Pezzo
Descrizione
- Lowest syringe carryover
- Reduced non specific sample interaction and increased analyte recovery
- Ultra smooth removable needles with choice of surface treatment.
Specifica
Specifica
| Include | Removable ultra smooth needle |
| Product Type | Diamond MS Manual Syringe |
| Modello | 10R-GT-MS1 |
| Quantità | Ea. |
| Volume (sistema metrico) | 10 μL |
| Tipo di collegamento | Removable |