Learn More
Trajan™ BP1 GC Capillary Columns
Suitable for analysis of hydrocarbons, aromatics, pesticides, phenol, herbicides, and amines. Trajan Scientific and Medical™ BP1 CG Capillary Columns are constructed of 100% Dimethyl Polysiloxane.
Marca: Trajan™ 054048
Dettagli aggiuntivi : Peso : 0.50000kg
Descrizione
Suitable for analysis of hydrocarbons, aromatics, pesticides, phenol, herbicides, and amines. Trajan Scientific and Medical™ BP1 CG Capillary Columns are constructed of 100% Dimethyl Polysiloxane.
- Classic cross-linked dimethyl polysiloxane technology
- Low bleed
- Non-polar
- Ideal for analyses such as hydrocarbons, aromatics, pesticides and amines
Specifica
0.22 mm | |
Van der Waals interaction | |
340/360°C | |
-60°C to +340/360°C | |
50 m | |
Non polar |
G1, G2, G38 | |
100% Dimethyl Polysiloxane | |
Amines Analysis, Aromatics Analysis, Pesticide Analysis, Phenols, Herbicides Analysis, Amines | |
0.25 μm | |
Forte™ BP1 | |
Fused Silica |