Learn More
SGE™ BPX-PCB Polycarborane-Siloxane GC Capillary Column
Click to view available options
Lunghezza (sistema metrico):
10 m
Dimensione della confezione:
Pezzo
Descrizione
Features
- Preferred column for polychlorinated biphenyl (PCB) compounds
- Separates PCB's on ortho ring substitution as well as boiling point
- Contains 8% Phenyl Polycarborane-siloxane
- Ideal for environmental analysis
- Unique high temperature phase suited for the analysis of persistent organic pollutants (POPs)
Applications
- Used for PCB congener analyses
- Nitro-substituted aromatics
- Polynuclear aromatic hydrocarbons
- Pesticides/herbicides
Specifica
Specifica
| Diametro (sistema metrico) interno | 0.1 mm |
| Stationary Phase | SGE™ BPX-PCB |
| Da utilizzare con (applicazione) | Polynuclear Aromatic Hydrocarbons Analysis, Nitro-substituted Aromatics, Polynuclear Aromatic Hydrocarbons, Pesticide Analysis, Herbicides Analysis |
| Dimensione particelle | 0.1 μm |
| Gamma di temperatura | -20°C to +360/370°C |
| Spessore pellicola | 0.1 μm |
| Lunghezza (sistema metrico) | 10 m |